missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
50 kg
6 x 1 kg
Unit Size:
1kg
50kg
5kg
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 kg |
| Linear Formula | K2SO4 |
| UN Number | NONH for all modes of transport |
| IUPAC Name | dipotassium;sulfate |
| Formula Weight | 174.26g/mol |
| Percent Purity | ≥99% |
| Grade | ACS/puriss. p.a. |
| Chemical Name or Material | Potassium Sulfate, Reag. ISO |
Safety and Handling
Product Identifier
- Potassium sulfate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction