missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfate, Puriss. p.a., ACS Reagent, Reag. ISO, ≥99%, Honeywell Fluka™

Product Code. 15683360 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
50 kg
6 x 1 kg
Unit Size:
1kg
50kg
5kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15683360

Brand: Honeywell-Fluka 312705KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. ISO,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate, dipotassium sulfate, potassium sulphate, sulfuric acid dipotassium salt, sal polychrestum, sulfuric acid, potassium salt, arcanum duplicatum, kalium sulphuricum, caswell no. 702, sulfuric acid, dipotassium salt
PubChem CID 24507
ChEBI CHEBI:32036
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Quantity 5 kg
Linear Formula K2SO4
UN Number NONH for all modes of transport
IUPAC Name dipotassium;sulfate
Formula Weight 174.26g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Sulfate, Reag. ISO
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.