missing translation for 'onlineSavingsMsg'
Learn More

Potassium sulfite, 90%, pure

Product Code. p-4813622 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass Bottle
Unit Size:
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 11914231

Brand: Thermo Scientific Acros 440210010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10117-38-1 | K2O3S | 158.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10117-38-1
Molecular Formula K2O3S
Molecular Weight (g/mol) 158.26
InChI Key BHZRJJOHZFYXTO-UHFFFAOYSA-L
Synonym potassium sulfite, dipotassium sulfite, potassium sulphite, sulfurous acid, dipotassium salt, unii-015kzc652e, sulfurous acid, potassium salt 1:2, potassium sulfite k2so3, kaliumsulfit, potassiumsulfite, dipotassium ion sulfite
PubChem CID 24958
IUPAC Name dipotassium;sulfite
SMILES [O-]S(=O)[O-].[K+].[K+]

Specifications

Quantity 1 kg
Assay Percent Range 88% min. (Iodimetry)
Linear Formula K2SO3
Merck Index 15,7795
Formula Weight 158.26
Percent Purity 90%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Potassium sulfite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.