missing translation for 'onlineSavingsMsg'
Learn More
Potassium sulfite, 90%, pure, Thermo Scientific Chemicals Product Code.: 11934231

Potassium sulfite, 90%, pure, Thermo Scientific Chemicals

Product Code. 11934231
250 g, Glass Bottle
Click to view available options
Quantity:
250 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 11934231

Brand: Thermo Scientific Acros 440212500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10117-38-1
Quantity 250 g
Assay Percent Range 88% min. (Iodimetry)
Molecular Formula K2O3S
Linear Formula K2SO3
Merck Index 15,7795
Synonym potassium sulfite, dipotassium sulfite, potassium sulphite, sulfurous acid, dipotassium salt, unii-015kzc652e, sulfurous acid, potassium salt 1:2, potassium sulfite k2so3, kaliumsulfit, potassiumsulfite, dipotassium ion sulfite
InChI Key BHZRJJOHZFYXTO-UHFFFAOYSA-L
SMILES [O-]S(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;sulfite
Molecular Weight (g/mol) 158.26
PubChem CID 24958
Formula Weight 158.26
Percent Purity 90%
Grade Pure
Packaging Glass Bottle
Chemical Name or Material Potassium sulfite
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464103 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog Number440210010,440210025,440212500
Lot NumberA0460111 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number440210010,440210025,440212500
Lot NumberA0440188 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number440210010,440210025,440212500
Lot NumberA0454043 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number440210010,440210025,440212500
Lot NumberA0449153 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number440210010,440210025,440212500
Product Identifier
  • Potassium sulfite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium sulfite, 90%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.