Learn More
Potassium tellurite monohydrate, 97%, Thermo Scientific Chemicals
£136.68 - £407.03
Chemical Identifiers
CAS | 123333-66-4 |
---|---|
Molecular Formula | K2O3Te·H2O |
MDL Number | MFCD00149923 |
Synonym | potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384947
|
Thermo Scientific Alfa Aesar
041959.14 |
25 g |
£136.68
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374947
|
Thermo Scientific Alfa Aesar
041959.22 |
100 g |
£407.03
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium tellurite, in association with agar is used as a selective medium for growth of clauberg (bacteria). It is used for the selective isolation of staphylococci and corynebacteria, which is used to reduce TeO3 2- to tellurium element.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123333-66-4 | |
K2O3Te·H2O | |
MFCD00149923 | |
potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e |
Specifications
465°C (decomposition) | |
White | |
97% | |
MFCD00149923 | |
Soluble in water. | |
White crystals |
123333-66-4 | |
Crystals | |
K2O3Te·H2O | |
potassium tellurite, potassium tellurate iv, dipotassium tellurite, dipotassium trioxotellurate, telluric acid, dipotassium salt, potassiium tellurite iv, telluric acid h2teo3 , dipotassium salt, dipotassium ion tellurite, acmc-1bm2e | |
253.80 Anhydrous | |
Potassium tellurite monohydrate |
Safety and Handling
P264b-P270-P301+P310-P330-P501c
H301
missing translation for 'dotInformation' : DOT Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 232-213-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only