Learn More
Potassium tert-butoxide, 1M solution in tert-butanol, AcroSeal™, Thermo Scientific Chemicals
£59.60 - £173.87
Chemical Identifiers
CAS | 865-47-4 |
---|---|
Molecular Formula | C4H9KO |
Molecular Weight (g/mol) | 112.21 |
MDL Number | MFCD00012162 |
InChI Key | LPNYRYFBWFDTMA-UHFFFAOYSA-N |
Synonym | potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok |
PubChem CID | 23665647 |
IUPAC Name | potassium;2-methylpropan-2-olate |
SMILES | CC(C)(C)[O-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10030564
|
Thermo Scientific Acros
426121000 |
100 mL | AcroSeal™ Glass Bottle |
£59.60
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10644555
|
Thermo Scientific Acros
426128000 |
800 mL | AcroSeal™ Glass bottle |
£173.87
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
865-47-4 | |
112.21 | |
LPNYRYFBWFDTMA-UHFFFAOYSA-N | |
23665647 | |
CC(C)(C)[O-].[K+] |
C4H9KO | |
MFCD00012162 | |
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok | |
potassium;2-methylpropan-2-olate |
Specifications
865-47-4 | |
(CH3)3COK | |
12°C | |
LPNYRYFBWFDTMA-UHFFFAOYSA-N | |
potassium;2-methylpropan-2-olate | |
23665647 | |
0.90 to 1.10M | |
0.82 | |
Potassium tert-butoxide, 1M solution in tert-butanol |
C4H9KO | |
MFCD00012162 | |
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok | |
CC(C)(C)[O-].[K+] | |
112.21 | |
112.21 | |
Colorless to amber solution | |
0.8200g/mL |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-740-3
- Potassium tert-butoxide, 1M solution in tert-butanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only