Learn More
Potassium tert-butoxide, pure, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 371221000
69.68 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
81.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Potassium tert-butoxide | |
Colorless to Orange-Yellow | |
100 mL | |
(CH3)3COK | |
−19°C | |
LPNYRYFBWFDTMA-UHFFFAOYSA-N | |
potassium;2-methylpropan-2-olate | |
23665647 | |
Pure | |
AcroSeal™ Glass bottle | |
0.9020g/mL |
865-47-4, 109-99-9 | |
Solution | |
C4H9KO | |
01,911; 02,336; 03,233; 04,399; 05,544; 06,477; 08,407; 09,380; 10,323; 11,432; 12,97; 14,264; 17,289 | |
potassium tert-butoxide, potassium tert-butanolate, potassium t-butoxide, potassium 2-methylpropan-2-olate, potassium tert-butylate, kotbu, 2-methyl-2-propanol, potassium salt, tert-butoxypotassium, potassium-t-butoxide, t-buok | |
CC(C)(C)[O-].[K+] | |
112.21 | |
112.21 | |
0.95 to 1.10M | |
0.902 |
Safety and Handling
- Potassium tert-butoxide, 1M solution in tetrahydrofuran
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458585 | Certificate of Analysis | 24/01/2024 | 371220010,371221000,371220025 |
A0448997 | Certificate of Analysis | 17/01/2024 | 371220010,371221000,371220025 |
A0453911 | Certificate of Analysis | 08/01/2024 | 371220010,371221000,371220025 |
A0394012 | Certificate of Analysis | 14/12/2022 | 371220010,371221000,371220025 |
A0413860 | Certificate of Analysis | 13/12/2022 | 371220010,371221000,371220025 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
6Minutes
3Seconds
53
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only