missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetraborate tetrahydrate, 99.5%, extra pure, Thermo Scientific Chemicals

Product Code. 10541301
Click to view available options
Quantity:
250 g
1 kg
5 kg
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 12045-78-2
Molecular Formula B4K2O7·4H2O
Molecular Weight (g/mol) 305.5
Synonym Potassium biborate tetrahydrate

Especificaciones

Melting Point 850.0°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.5%
Linear Formula K2B4O7·4H2O
Merck Index 14,768
Solubility Information Solubility in water: soluble.
Formula Weight 305.5
Percent Purity 99.5%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Potassium tetraborate tetrahydrate
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'