missing translation for 'onlineSavingsMsg'
Learn More

Potassium Tetraborate Tetrahydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10040310 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10040310

Brand: Fisher Chemical P/7140/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 850°C
CAS 12045-78-2
Color White
pH 9.2 to 9.3
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula B4K2O7·4H2O
MDL Number 149924
Formula Weight 305.5g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Potassium tetraborate tetrahydrate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.