missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrachloroaurate(III), 98%

Product Code. 10677831 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10677831

Brand: Thermo Scientific Acros 316120010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13682-61-6 | AuCl4K | 377.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13682-61-6
Molecular Formula AuCl4K
Molecular Weight (g/mol) 377.88
MDL Number MFCD00043088
InChI Key JMONCQYBAMKVQQ-UHFFFAOYSA-J
Synonym potassium tetrachloroaurate iii, potassium tetrachloroaurate, k aucl4, kaucl4 * 2h2o, potassium tetrachloridoaurate, potassium ion tetrachloroaurate, potassium tetrachloroaurate 1-, potassium tetrachloridoaurate 1-, potassium tetrachloridoaurate iii, potassium gold iii chloride
PubChem CID 11337898
ChEBI CHEBI:51564
IUPAC Name potassium;tetrachlorogold(1-)
SMILES Cl[Au-](Cl)(Cl)Cl.[K+]

Specifications

Melting Point >300.0°C
Color Yellow to Orange
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 97.5% min
Linear Formula KAuCl4
Merck Index 15, 7800
Solubility Information Solubility in water: soluble
Formula Weight 377.88
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Potassium tetrachloroaurate(III)
Show More Show Less
compliance-icons
Product Identifier
  • Potassium tetrachloroaurate(III)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt