Learn More
Potassium tetrachloroplatinate(II), 46-47% Pt, Thermo Scientific Chemicals
£104.52 - £1426.10
Chemical Identifiers
CAS | 10025-99-7 |
---|---|
Molecular Formula | Cl4H4K2Pt |
Molecular Weight (g/mol) | 419.11 |
MDL Number | MFCD00011378 |
InChI Key | JCAXKWJYKUEOFO-UHFFFAOYSA-N |
Synonym | potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate |
PubChem CID | 61440 |
SMILES | Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10594131
|
Thermo Scientific Acros
195360010 |
1 g | Glass bottle |
£104.52
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10144260
|
Thermo Scientific Acros
195360050 |
5 g | Glass bottle |
£372.86
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10348753
|
Thermo Scientific Acros
195360250 |
25 g | Glass bottle |
£1426.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-99-7 | |
419.11 | |
JCAXKWJYKUEOFO-UHFFFAOYSA-N | |
61440 |
Cl4H4K2Pt | |
MFCD00011378 | |
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate | |
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] |
Specifications
250.0°C | |
Red to Brown | |
Cl4H4K2Pt | |
MFCD00011378 | |
15, 7801 | |
Solubility in water: 10g/L (20°C) | |
Cl.Cl.Cl.Cl.[K+].[K+].[Pt--] | |
419.11 | |
415.1 | |
Potassium tetrachloroplatinate(II) |
10025-99-7 | |
Crystalline Powder and/or Crystals | |
K2PtCl4 | |
02,182; 04,215 | |
potassium tetrachloroplatinate ii, dipotassium tetrachloroplatinate, potassium tetrachloroplatinate, potassium chloroplatinite, potassium tetrachloroplatinite, dipotassium tetrachloroplatinate 2-, potassium platinochloride, platinous potassium chloride, potassium platinous chloride, bipotassium tetrachloroplatinate | |
JCAXKWJYKUEOFO-UHFFFAOYSA-N | |
dipotassium tetrachloroplatinumdiuide | |
61440 | |
46 to 47% Pt |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-050-9
- Potassium tetrachloroplatinate(II)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Potassium tetrachloroplatinate(II)
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.