missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium tetranitroplatinate(II), Pt 42.6% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Potassium tetranitroplatinate(II) is used widely as laboratory chemicals and in manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Crystalline |
Quantity | 1 g |
Solubility Information | It is slightly soluble in water. It is soluble in organic or non-aqueous solvent. |
IUPAC Name | dipotassium;platinum;tetranitrite |
Formula Weight | 457.32 |
Concentration or Composition (by Analyte or Components) | Pt 42.6% min. |
Chemical Name or Material | Potassium tetranitroplatinate(II) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'