missing translation for 'onlineSavingsMsg'
Learn More

Potassium thiocyanate, 0.1N Standardized Solution, Thermo Scientific Chemicals

£44.42 - £94.13

Chemical Identifiers

CAS 333-20-0
Molecular Formula CKNS
Molecular Weight (g/mol) 97.176
MDL Number MFCD00011413
InChI Key ZNNZYHKDIALBAK-UHFFFAOYSA-M
Synonym potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
PubChem CID 516872
ChEBI CHEBI:30951
IUPAC Name potassium;thiocyanate
SMILES C(#N)[S-].[K+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393707
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035602.K2
1 L
£44.42
1 L
Please to purchase this item. Need a web account? Register with us today!
11383707
View Documents
Thermo Scientific Alfa Aesar
035602.K7
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Potassium thiocyanate, 0.1N standardized solution is used in analytical chemistry for the quantitative determination of Fe(III). In Volhard’s method, it is used to determine the concentration of chloride ion by titration. It is also used as an analyzer for direct and continuous measurement of the concentration of soluble copper in an aqueous solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

333-20-0
97.176
ZNNZYHKDIALBAK-UHFFFAOYSA-M
516872
potassium;thiocyanate
CKNS
MFCD00011413
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
CHEBI:30951
C(#N)[S-].[K+]

Specifications

333-20-0
Liquid
MFCD00011413
Miscible with water,acetone and alcohol.
C(#N)[S-].[K+]
97.176
CHEBI:30951
Odorless
23 hPa (17mm Hg) at 20°C
Colorless
CKNS
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt
ZNNZYHKDIALBAK-UHFFFAOYSA-M
potassium;thiocyanate
516872
97.18
Colorless liquid
Potassium thiocyanate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 206-370-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only