Learn More
Potassium thiocyanate, 99+%, for analysis, Thermo Scientific Chemicals
£33.40 - £528.00
Chemical Identifiers
CAS | 333-20-0 |
---|---|
Molecular Formula | CKNS |
Molecular Weight (g/mol) | 97.18 |
MDL Number | MFCD00011413 |
InChI Key | ZNNZYHKDIALBAK-UHFFFAOYSA-M |
Synonym | potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt |
PubChem CID | 516872 |
ChEBI | CHEBI:30951 |
IUPAC Name | potassium;thiocyanate |
SMILES | C(#N)[S-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10086063
|
Thermo Scientific Acros
196582500 |
250 g | Plastic bottle |
£33.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10002490
|
Thermo Scientific Acros
196585000 |
500 g | Plastic bottle |
£59.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10562863
|
Thermo Scientific Acros
196580025 |
2.5 kg | Plastic bottle |
£178.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10618313
|
Thermo Scientific Acros
196580100 |
10 kg | Plastic drum |
£528.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
333-20-0 | |
97.18 | |
ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
516872 | |
potassium;thiocyanate |
CKNS | |
MFCD00011413 | |
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
CHEBI:30951 | |
C(#N)[S-].[K+] |
Specifications
170.0°C to 179.0°C | |
White | |
Crystalline Powder | |
CKNS | |
MFCD00011413 | |
15, 7807 | |
Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone | |
C(#N)[S-].[K+] | |
97.18 | |
CHEBI:30951 | |
≥99% | |
1.886 | |
Potassium thiocyanate, p.a. |
333-20-0 | |
5.3 to 8.7 (5% soln. at 20°C) | |
99% min. (Argentometry) | |
KSCN | |
01,954 | |
potassium thiocyanate, potassium rhodanate, potassium rhodanide, rodanca, potassium sulfocyanate, rhodanide, rhocya, potassium thiocyanide, kscn, thiocyanic acid, potassium salt | |
ZNNZYHKDIALBAK-UHFFFAOYSA-M | |
potassium;thiocyanate | |
516872 | |
97.18 | |
Analytical | |
1.8860g/mL |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-370-1
missing translation for 'rtecsNumber' : XL1925000
missing translation for 'tsca' : TSCA
- Potassium thiocyanate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P233-Keep container tightly closed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only