missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Thiocyanate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless or White |
Quantity | 2.5 kg |
Linear Formula | KSCN |
UN Number | NONH for all modes of transport |
Beilstein | 3594799 |
Formula Weight | 97.18g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Chemical Name or Material | Potassium Thiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'