missing translation for 'onlineSavingsMsg'
Learn More

Potassium thioglycolate, 40-44 wt.% solution in water, Thermo Scientific™

Product Code. 10509993
Click to view available options
Quantity:
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34452-51-2, 7732-18-5
Molecular Formula C2H3KO2S
Molecular Weight (g/mol) 130.202
MDL Number MFCD00067265
InChI Key HYTYHTSMCRDHIM-UHFFFAOYSA-M
Synonym potassium thioglycolate, unii-xp5wzo5w6h, potassium 2-mercaptoacetate, acetic acid, mercapto-, monopotassium salt, xp5wzo5w6h, 68-11-1 parent, potassium sulfanylacetate, potassium mercaptoacetate, kopcolate, potassium ion thioglycolate
PubChem CID 23688957
IUPAC Name potassium;2-sulfanylacetate
SMILES C(C(=O)[O-])S.[K+]

Specifications

Color Pink
Density 1.2500g/mL
Solubility Information Solubility in water: soluble.
Formula Weight 130.21
Percent Purity 40.0 to 44.0%
Identification potassium: positive, mercaptane: positive
Packaging Glass bottle
Specific Gravity 1.25
pH 6.8 to 7.3
Quantity 250 mL
Physical Form Solution
Chemical Name or Material Potassium thioglycolate, 40-44 wt.% solution in water
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium thioglycolate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H301-Toxic if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'