Learn More
Potassium thioglycolate, 40-44 wt.% solution in water, Thermo Scientific™
£12.05
Chemical Identifiers
CAS | 34452-51-2, 7732-18-5 |
---|---|
Molecular Formula | C2H3KO2S |
Molecular Weight (g/mol) | 130.202 |
MDL Number | MFCD00067265 |
InChI Key | HYTYHTSMCRDHIM-UHFFFAOYSA-M |
Synonym | potassium thioglycolate, unii-xp5wzo5w6h, potassium 2-mercaptoacetate, acetic acid, mercapto-, monopotassium salt, xp5wzo5w6h, 68-11-1 parent, potassium sulfanylacetate, potassium mercaptoacetate, kopcolate, potassium ion thioglycolate |
PubChem CID | 23688957 |
IUPAC Name | potassium;2-sulfanylacetate |
SMILES | C(C(=O)[O-])S.[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10509993
|
Thermo Scientific Acros
384702500 |
250 mL | Glass bottle | N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
34452-51-2, 7732-18-5 | |
130.202 | |
HYTYHTSMCRDHIM-UHFFFAOYSA-M | |
23688957 | |
C(C(=O)[O-])S.[K+] |
C2H3KO2S | |
MFCD00067265 | |
potassium thioglycolate, unii-xp5wzo5w6h, potassium 2-mercaptoacetate, acetic acid, mercapto-, monopotassium salt, xp5wzo5w6h, 68-11-1 parent, potassium sulfanylacetate, potassium mercaptoacetate, kopcolate, potassium ion thioglycolate | |
potassium;2-sulfanylacetate |
Specifications
Pink | |
C2H3KO2S | |
1.2500g/mL | |
Solubility in water: soluble. | |
C(C(=O)[O-])S.[K+] | |
130.202 | |
130.21 | |
potassium: positive, mercaptane: positive | |
6.8 to 7.3 | |
Potassium thioglycolate, 40-44 wt.% solution in water |
34452-51-2, 7732-18-5 | |
MFCD00067265 | |
potassium thioglycolate, unii-xp5wzo5w6h, potassium 2-mercaptoacetate, acetic acid, mercapto-, monopotassium salt, xp5wzo5w6h, 68-11-1 parent, potassium sulfanylacetate, potassium mercaptoacetate, kopcolate, potassium ion thioglycolate | |
HYTYHTSMCRDHIM-UHFFFAOYSA-M | |
potassium;2-sulfanylacetate | |
23688957 | |
40.0 to 44.0% | |
1.25 | |
Solution |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Potassium thioglycolate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H301-Toxic if swallowed.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.