missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium trimethylacetate, 95%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Potassium trimethylacetate is used as a intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium trimethylacetate is used as a intermediate for chemical research.
Notes
Keep container tightly closed. Store away from oxidizing agent.
Potassium trimethylacetate is used as a intermediate for chemical research.
Notes
Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | ∼200°C |
| Linear Formula | (CH3)3CCO2K |
| Quantity | 25 g |
| Formula Weight | 140.22 |
| Percent Purity | 95% |
| Chemical Name or Material | Potassium trimethylacetate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction