missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UF30
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Brand: Memmert™ UF30H6
This item is not returnable.
View return policy
Description
- Accessory
- For combination error message
Specifications
UF30 Oven | |
Socket to NAMUR NE 28 | |
2 A |
Potential-free Contact | |
24 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UF30 > For Use With: UF30
Spot an opportunity for improvement?Share a Content Correction