missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA556895
This item is not returnable.
View return policy
Description
Immunogen sequence: RSPHVAHHSP HTNHPNAWGA SPAPNPSITS SGQPLNVYSQ PGFTVSGMLE HGGLTPPPAA ASAQSLHPVL REPPDHGELG SHHCQDH Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.Specifications
POU3F4 | |
Polyclonal | |
Unconjugated | |
POU3F4 | |
brain-4; brain-4 class III POU-domain protein; brain-specific homeobox/POU domain protein 4; BRN4; Brn-4; DFN3; DFNX2; OCT-9; Octamer-binding protein 9; octamer-binding transcription factor 9; Otf9; OTF-9; POU class 3 homeobox 4; POU domain, class 3, transcription factor 4; POU3F4; RHS2; RHS2 class III POU protein; sex linked fidget; Slf | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5456 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49335 | |
POU3F4 | |
Recombinant protein corresponding to Human POU3F4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only