missing translation for 'onlineSavingsMsg'
Learn More

Pouds™ Wall Brackets

For attaching sharps containers to the wall

£3.90

Specifications

Color Red
For Use With (Application) Attaching sharps container to a wall
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
12993505
View Documents
Daniels Healthcare
DD487
Sharpsguard mini, 1, 2.5, 3.75, 5 and com plus
£3.90
Each
Please to purchase this item. Need a web account? Register with us today!
12903515
View Documents
Daniels Healthcare
DD488
Sharpsguard 7, 11.5, and 22
£3.90
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Wall bracket for SHARPSGUARD™ mini, 1, 2.5, 3.75, 5 and com plus containers. Red.
Specifications

Specifications

Red
Attaching sharps container to a wall
Videos
SDS
Documents

Documents

Product Certifications