Learn More
Pour Point Standard, Paragon Scientific™
Pour Point Certified Reference Materials for the verification of laboratory instruments used in testing petroleum and derivative products for Pour Point to IP 15, ASTM D97, NFT 60-105.
Brand:  Paragon Scientific CRMU-PPLU
Product Code. 15866862
  • £244.00 / 250mL
Explore more special offers
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom
Specifications
Specifications
250 mL
Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pour Point Standard, Paragon Scientific™ > Quantity: 250mL
Nominal values quoted