Learn More
Pour Point Standard IP15, ASTM D97, ISO 3016, BS 2000, Pt 15 (-30.5 °C) UKAS ISO 17034 Certified 250mL, Paragon Scientific™
Pour Point Certified Reference Materials for the verification of laboratory instruments used in testing petroleum and derivative products for Pour Point to IP 15, ASTM D97, NFT 60-105.
Brand: Paragon Scientific CRMU-PPGO
Additional Details : CAS Number : 68334-30-5 Weight : 0.20000kg Transport : UN number : 1202 Chem class : 3 Pack group : III
Description
- Certified according to ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Certifications
Nominal values quoted
Specifications
UN 1202 |
Safety and Handling
- CRMU-PPGO
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Aspiration toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
Nominal values quoted. Certified Values will vary from batch to batch.