Learn More
Labcold™ Powder Coated Steel Pharmacy Refrigerators
Brand:  Labcold™ RPFR21043
 View more versions of this product
Product Code. 12928614
  • £14730.00 / Each
Explore more special offers
Description
Description
  • Microprocessor controlled temperature regulation for accuracy and stability
  • Temperature range +2°C to +8°C
  • Fan circulated interior
  • Stainless steel construction
  • Door locks fitted as standard
  • Digital temperature display with audio alarm
  • Heavy duty lockable castors on larger models
  • Optimised air circulation
Specifications
Specifications
820 x 720 x 1980mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labcold™ Powder Coated Steel Pharmacy Refrigerators > Exterior Dimensions (H x W x D): 1980 x 720 x 820mm