Learn More
Hanna Instruments™ Free Chlorine Reagents (100 tests)
High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements.
Brand: Hanna Instruments™ HI9370101
This item is not returnable.
View return policy
Description
- Reagents for the colorimetric determination of free chlorine
- There are enough reagents for 100 tests and are for use with Hanna’s compatible benchtop and portable photometers
- High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
- Follow the DPD Method in which the reaction between chlorine and reagent causes a pink tint in the sample
- Designed to be used with samples that have an expected range of 0.00 to 5.00 mg/L free chlorine
Specifications
100 Pack |