missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Usbeck™ Powder Spatula
Transfer substances with simplicity. Usbeck™ Powder Spatula is ideal for transferring substances.
Brand: Usbeck™ 3282
Description
- Stainless steel
- Width of blade 9 mm
Specifications
Powder Spatula | |
Stainless Steel | |
150 mm |