Learn More
Invitrogen™ Power Blotter Cassette
Get rapid and efficient semi-dry protein transfer with the Invitrogen™ Power Blotter.
Brand: Invitrogen™ PB0002
Includes: Invitrogen Power Blotter Cassette
Description
The Invitrogen™ Power Blotter Cassette is part of the Power Blotter System designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes.
This item is made available as a replacement or additional cassette for the Power Blotter Station.
The Power Blotter Cassette can be used with Power Blotter Select Transfer Stacks for rapid transfers, the Power Blotter 1-Step Transfer Buffer for semi-dry transfers, or alternatively with traditional buffers for longer semi-dry transfers.
Specifications
Western Blot | |
Nitrocellulose, PVDF | |
Power Blotter | |
Blotter Cassette | |
Room Temperature |
PB0010 Power Blotter Station | |
Semi-Dry | |
1 Midi or 2 Mini Gels | |
1 cassette |