Learn More
Edulab™ Power Supply Eht
Brand:  Edulab™ 605-006
Product Code. 12826472
  • £400.00 / Each
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
adjustable 5KV with digital display, provides EHT outputs for electrostatic experiments, with short circuit current of 60μA 110-230V 50/60Hz blue 208mm x 155mm x 115mm 0KV to 5KV DC, 2mA Edu-Lab