missing translation for 'onlineSavingsMsg'
Learn More

UVP Power supply Pen-Ray PS2 RMS 2000 Vac lamp current 6mA 220-240V 50/60Hz for use with Pen-Ray lamp UVP

Brand:  UVP 99-0056-02

Product Code. 12823828

  • £336.00 / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Pen-Raylamp
220/240 V
Power supply for Pen-Ray PS2 RMS 2000 Vac lamp
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
UVP Power supply Pen-Ray PS2 RMS 2000 Vac lamp current 6mA 220-240V 50/60Hz for use with Pen-Ray lamp UVP >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.