Learn More
Invitrogen™ PowerEase™ Touch 350W Power Supply and Mini Gel Tank Welcome Pack
The PowerEase Touch 350W Power Supply and Mini Gel Tank Welcome Pack contains: PowerEase Touch 350W Power Supply and Mini Gel Tank
Brand: Invitrogen™ PSC350M
Description
The PowerEase Touch 350W Power Supply and Mini Gel Tank Welcome Pack contains:
- PowerEase Touch 350W Power Supply
- Mini Gel Tank
About the PowerEase Touch 350W Power Supply
The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.
About the Mini Gel Tank
The Mini Gel Tank is compatible with all Invitrogen Novex, NuPAGE, and Bolt mini gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient, side-by-side gel loading and enhanced viewing during use. Run times may vary depending on buffer conditions and power supplies used.
Specifications
Room Temperature | |
Mini | |
PowerEase | |
4.3 inch backlit LCD | |
1 pack | |
Yes | |
30.3 inch x 23 inch x 10.2 inch | |
300 VDC |
High throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE and western blotting | |
3 A | |
Up to 999 min./step | |
4 (4 positive voltage and 4 negative voltage) | |
Room Temperature | |
One year | |
2.1 kg | |
350 watts |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.