Learn More
3M™ Powered Air Respirator System Starter Kit
Powered Air Respirator System Starter Kit is a durable powered air respirator system designed for heavy duty environments at an affordable price. This starter kit includes a turbo unit, battery, charger and belt.
Brand:  3M™ 7100299964
748.19 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 17850877
  • £1115.00 / Each
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For a durable, economical powered air respirator starter kit use 3M™ Powered Air Respirator System Starter Kit PF-619E.
  • This kit includes a 3M™ PF-600E turbo unit, charger and belt.
  • The 3M™ PF-600E turbo unit features audible and visual alarms that alert the user to low battery or if the flow rate falls below the manufacturer's minimum design flow rate of 160 l/min.
  • The unit incorporates a simple button operation, and the button is shielded to prevent accidental shut-off.
  • It's designed to be compatible with loose fitting 3M™Versaflo™ M-200 and M-300 range of helmets, tight fitting 3M™ Promask™ FM3 and 3M™ Vision FM4 Full facepieces and the 3M™ DT-Series range of filters.
  • The unit has an IP 65 rating with filters and decontamination plugs fitted to the filter and hose ports, making it suitable for cleaning by showering/water spray.
  • The integrated battery is safely enclosed within the turbo unit casing and is rechargeable in-situ.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Various
Standard
Reusable
PAPR Kit