missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-100 Heavy Duty Crinkle Latex Coated Gloves

Product Code. 11736673
Click to view available options
Size:
7
8
9
10
11

Automatic knit construction delivers an optimally tensioned inner surface for superb fit and flexibility. Latex coating features a raised crinkle finish for enhanced grip in wet and dry conditions

  • Gloves deliver overall mechanical performance for maximal protection and durability
  • Latex coating's raised crinkle finish enhances grip in both dry and wet conditions
  • Automatic knitted liner delivers an optimally tensioned inner surface for improved fit and flexibility
  • Designed for use in a wide range of handling and carrying applications
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Cuff Material Latex
Coating Material Crinkled Latex
Color Gray, Blue
Cut Resistance Rating ANSI A2
Exterior Finish Partially Textured
Hand Specific
Material Polycotton
Product Type Dip-coated Gloves
Size 11
For Use With (Application) Machinery and Equipment
Glove Cuff Style Knit
Series 80-100
Yarn Gauge 10
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'