missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-100 Heavy Duty Crinkle Latex Coated Gloves

Product Code. 11756673
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Automatic knit construction delivers an optimally tensioned inner surface for superb fit and flexibility. Latex coating features a raised crinkle finish for enhanced grip in wet and dry conditions

  • Gloves deliver overall mechanical performance for maximal protection and durability
  • Latex coating's raised crinkle finish enhances grip in both dry and wet conditions
  • Automatic knitted liner delivers an optimally tensioned inner surface for improved fit and flexibility
  • Designed for use in a wide range of handling and carrying applications
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Cuff Material Latex
Coating Material Crinkled Latex
Color Gray, Blue
Cut Resistance Rating ANSI A2
Exterior Finish Partially Textured
Hand Specific
Material Polycotton
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Machinery and Equipment
Glove Cuff Style Knit
Series 80-100
Yarn Gauge 10
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'