missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ PowerFlex™ 80-658 Series Aramid Fiber Heavy Weight Gloves

Product Code. 15646906
Click to view available options
Size:
6
7
8
9
10
11

Protects wearer with triple-layer construction comprised of steel, glass fiber and aramid fiber provides extreme resistance to cuts, splinters and burrs.

  • Mechanical protection gloves
  • Exceeds EN388 cut level 5.
  • Extra-long cuff offers extended safety for the forearm.
  • ¾ dipped and liquid repellent, shielding the skin from unpleasant liquid solutions.
  • Good heat contact resistance.
  • EN407 level-2 rating means safe handling of warm objects.
  • Natural rubber crinkle finish provides a firm grip.
  • Long life provides a highly cost-effective solution.
  • Heather green color helps mask dirt.
  • Excellent washability, with no decrease in comfort or fading.

Applications

  • Handling heavy loads, metal edges
  • Waste handling: recycling, sorting (glass, cans, metal pieces)
  • Handling glass sheets, sharp-edged objects
  • Handling metal sheets, cutting of dry, painted or galvanized pieces
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 407:2004, EN 388:2003
Coating Material Latex
Color Blue, Green
Exterior Finish 3/4 Dipped
Latex Free No
Material Dupont™ Kevlar™ Aramid Fibers
Product Type Cut-resistant Gloves
Size 7
Style Construction: Knitted
Glove Cuff Style Long Knitwrist
Liner Material Stainless steel, Nylon, Glass fiber, Cotton, Dupont™ Kevlar™ aramid fiber
Packaging Type 12 pairs in a bag, 6 bags in a carton
Series 80-658
Length (Metric) 260 to 295 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'