missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PowerTrack™ SYBR Green Master Mix for qPCR

Product Code. 16505231 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 mL
10 x 5 mL
2 x 5 mL
5 mL
5 x 5 mL
50 mL
Unit Size:
10mL
1mL
25mL
50mL
5mL
This item is not returnable. View return policy

Product Code. 16505231

Brand: Applied Biosystems™ A46012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PowerTrack SYBR Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows with built-in visual dye–based indicators for accurate reaction set up.

PowerTrack SYBR Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows with built-in visual dye–based indicators for accurate reaction set up. Coupled with user-supplied primer sets and template, PowerTrack SYBR Green Master Mix is designed to amplifiy targets for accurate gene expression analysis.

Features of PowerTrack SYBR Green Master Mix include:

  • Built-in visual indicators aid in reaction setup
  • Broad primer Tm and concentration compatibility allows qPCR reaction setup flexibility with minimal optimization
  • Specific and tight reproducibility in Ct's over a broad dynamic range
  • Formulated with UNG/dUTP to prevent contamination of carryover PCR products

Built-in visual indicators

PowerTrack SYBR Green Master Mix contains an inert blue dye that does not interfere with real-time PCR but provides increased visibility in the tube or well. A separate sample buffer is provided that contains an inert yellow dye. The optional use of this yellow sample buffer results in a reaction mixture that changes from blue to green upon addition of the sample, helping one avoid pipetting mistakes. The yellow sample buffer aids in reaction setup, but is not required to obtain superior results with PowerTrack SYBR Green Master Mix.

Formulated for maximum specificity and reproducibility

PowerTrack SYBR Green Master Mix contains Taq DNA polymerase, which is tightly controlled through an antibody-mediated hot-start mechanism, helping prevent undesirable early activity of the polymerase at low temperatures that can lead to nonspecific amplification. PowerTrack SYBR Green Master Mix is designed for exceptional performance in your most challenging real-time PCR applications.

Broad instrument compatibility

PowerTrack SYBR Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems real-time PCR instruments. It is also compatible with the Bio-Rad CFX96™, CFX384™, and iQ™5; Roche LightCycler™ 480; and Agilent Mx3005P™ systems. For optimal results, primer concentrations should be in the range of 300–800 nM.

UNG for carryover contamination control

Contamination in labs that routinely run PCR is a major concern for many researchers and is the source of most false positives. The inclusion of UNG and dUTP in PowerTrack SYBR Green Master Mix results in the degradation of any previously amplified PCR products and helps prevent contamination of subsequent qPCR reactions.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • PowerTrack SYBR Green Master Mix
• 40X Yellow Sample Buffer

Store at -20±5°C in the dark.
Detection Method SYBR
Format Tube
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Flex, QuantStudio 7 Pro, QuantStudio 12K Flex, Bio-Rad CFX Connect, Bio-Rad CFX384 Touch, Bio-Rad CFX96 Touch, BioRad CFX96, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, ViiA 7 System, StepOne System, MJ Chromo4, MJ Opticon, StepOnePlus System, Roche LightCycler 480, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000
Multiplex Capability Single-plex
Passive Reference Dye ROX (Pre-mixed)
Product Line Applied Biosystems
Product Type Real Time PCR SYBR Master Mix
Quantity 1 mL
Shipping Condition Dry Ice
Sufficient For 100 Reactions
Target Specificity cDNA or gDNA only
For Use With (Application) Gene Expression, Microbial Detection
Thermostability 8 hr., Assembled Reaction
No. of Reactions 100 Reactions
Sample Type DNA
Volume 1 mL
Includes dUTP, UNG/UDG
Inhibitor Tolerance (Simple) High
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis