missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PowerUp™ SYBR™ Green Master Mix for qPCR

Product Code. 15320929 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 mL
5 mL
2 x 1 mL
2 x 5 mL
50 mL
5 x 1 mL
5 x 5 mL
10 x 1 mL
10 x 5 mL
Unit Size:
1mL
50mL
5mL
Pack of 10
Pack of 2
Pack of 5
This item is not returnable. View return policy

Product Code. 15320929

missing translation for 'mfr': Applied Biosystems™ A25741

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-formulated, optimized, universal 2X master mix for real-time PCR workflows.

PowerUp™ SYBR™ Green Master Mix is a pre-formulated, optimized, universal 2X master mix for real-time PCR workflows. Coupled with user-supplied primer sets and template, PowerUp™ SYBR™ Green Master Mix is designed to amplifiy targets for accurate gene expression analysis.

PowerUp™ SYBR™ Green Master Mix features include:

  • A dual hot-start mechanism for excellent specificity
  • Highly reproducible CTs over a broad dynamic range
  • Inclusion of UDG to help prevent carryover contamination
  • Stability of pre-assembled reactions for up to 72 hours
  • Compatibility with most real-time qPCR instruments

Exceptional specificity with dual hot-start mechanism

Specificity is paramount in obtaining high-quality data in SYBR™ Green reactions and can be enhanced through control of the Taq DNA polymerase at lower temperatures before stringent primer binding. PowerUp™ SYBR™ Green Master Mix uses the Dual-Lock™ Taq DNA polymerase enzyme that combines two unique hot-start mechanisms to help control its activity and to prevent undesirable early activity of the polymerase at low temperatures. In an experiment evaluating 24 different primer sets, PowerUp™ SYBR™ Green Master Mix generated single melt curves 100% of the time, consistent with highly specific amplification.

Tight reproducibility in CTs over a wide dynamic range

PowerUp™ SYBR™ Green Master Mix demonstrates excellent reproducibility over a wide dynamic range for a variety of targets tested (see figure). In addition, this master mix provides efficient amplification over 6 logs of sample input. Unlike competitor mixes that can exhibit inhibition at high cDNA inputs and low correlation coefficients over a dynamic range experiment, PowerUp™ SYBR™ Green Master Mix efficiency is maintained over the full dynamic range to help deliver maximal confidence in data quality (see figure).

UDG included for carryover contamination control

The inclusion of uracil-DNA glycosylase (UDG) and dUTP in the PowerUp™ SYBR™ Green Master Mix enables the degradation of any previously amplified PCR products, helping prevent contamination of subsequent qPCR reactions and possible false positives.

Stability of pre-assembled reactions

The tight control of the Dual-Lock™ Taq DNA polymerase in PowerUp™ SYBR™ Green Master Mix enables reactions to be assembled up to 72 hours prior to cycling without impacting data (see figure). This consistency allows users to have confidence and flexibility in the use of PowerUP™ SYBR™ Green Master Mix across numerous workflow scenarios¹.

Broad instrument compatibility

PowerUp™ SYBR™ Green Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments². It is also compatible with the Bio-Rad IQ™5 and CFX96™ / CFX384™ systems, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems. For optimal results, recommended primer concentrations are 300–800 nM.

¹Pre-PCR stability is influenced not only by the master mix, but also by the target being analyzed. For maximum confidence, researchers should confirm stability profiles of their specific targets.

² For optimal performance, standard cycling using the Applied Biosystems™ 7900 system is recommended.

Order Info

Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage The 2X mix contains SYBR Green dye, Dual-Lock Taq DNA Polymerase, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye, and optimized buffer components. Contains 1 x 1 mL tube, sufficient for 100 20-μL reactions (10 μL Master Mix per reaction).

Store at 2–8°C in the dark.
Detection Method SYBR
Format Tube
PCR Method qPCR
Polymerase Dual-Lock Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System, 7900HT System, QuantStudio 12k Flex, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio 7 Flex, StepOne, StepOnePlus, ViiA 7 System
Multiplex Capability Single-plex
Passive Reference Dye ROX (Pre-mixed)
Product Line PowerUP, SYBR
Product Type Real Time PCR SYBR Master Mix
Quantity 1 mL
Shipping Condition Wet Ice
Sufficient For 100 Reactions of 20 μL
Target Specificity cDNA or gDNA only
For Use With (Application) Gene Expression
Thermostability 72 hr., Assembled Reaction
No. of Reactions 100 Reactions
Sample Type DNA
Volume 1 mL
Includes dUTP, UNG/UDG
Inhibitor Tolerance (Simple) Medium
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.