missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Poxygrid™ Biohazard Bag Holders

Product Code. p-31547 Shop All SP Scienceware Products
Click to view available options
Diameter (English):
7.3 in.
14.6 in.
Height (English):
20.5 in.
23.5 in.
29.5 in.
Unit Size:
Each

Product Code. 12370619

Brand: Bel-Art™ H131890000

Only null left
Add to basket
Add to basket

Sturdy wire holder keeps bags open while a separate hooked ring that rests over the top keeps them securely in place

  • Bright biohazard orange colored epoxy-coated steel wire
  • Chemical and corrosion-resistant; unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F)
  • Autoclavable

Specifications

Description Circular epoxy coated wire frame with easy-to-use hooked ring over a top
Autoclavable Autoclavable
Color Orange
Disposable Reusable
For Use With (Application) To keep biohazard bags open and ready for use
For Use With (Equipment) 01-814-3, 01-814C, 01-815C, 01-826-7, 01-826C, 01-827D, 01-828D, 01-828-033, 01-829D, 01-831A, 01-832A 36 H x 24 in. W biohazard bags
Material Epoxy-coated Wire
Temperature Range (English) -369°F to +250°F
Temperature Range (Metric) -223°C to +121°C
Width (English) 14.6 in.
Width (Metric) 37 cm
Height (English) 29.5 in.
Height (Metric) 75 cm
Diameter (English) 14.6 in.
Diameter (Metric) 37 cm
Product Type Biohazard Bag Holder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.