missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Poxygrid Lead Flask Ring Holder
Wire formed, epoxy coated holder keeps lead ring weights organized and handy
Brand: Bel-Art™ SP Scienceware™ F18307-1000
Code : 03
Additional Details : Weight : 0.10800kg
Description
- Small footprint; circular base is only 19cm (7½ in.) in diameter
- Rings having an inside diameter of 17.8mm (0.7 in.) or larger stack neatly on a tapered center post (17.1cm (6¾ in.) high)
- Steam autoclavable at 121°C (250°F)
Specifications
Blue | |
Epoxy Coated Wire | |
1 |
19 cm | |
17.1 cm | |
Autoclavable |