missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Poxygrid Steel Test Tube Racks

Chemical and corrosion resistant steel test tube racks in different colors for easy identification.

Brand:  Bel-Art™ SP Scienceware™ F18772-0000

 View more versions of this product

Product Code. 15570032

  • £51.65 / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Chemical and corrosion resistant steel test tube racks in different colors for easy identification.

  • Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy
  • Orange racks can be used for biohazard warning, yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of −223°C (−369°F) to 121°C (250°F)
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications

Specifications

Test Tube Rack
Autoclavable
Tubes (20 to 25 mm)
40
Non-sterile
-223°C to +121°C
25 mm
4 x 10
Green
Epoxy-coated Steel
Rectangular
20 - 25mm Tubes
29.3 x 12.8 x 8.3 cm (11.56 x 5.06 x 3.25 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers