missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ Polypropylene, Stopcock
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Specifications
Specifications
| Closure Type | Stopcock |
| Color | Blue |
| For Use With (Equipment) | For Carboy Series 350 |
| Material | Polypropylene |
| No. per Pack | 1 Ea. |
| Product Type | Stopcock |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction