missing translation for 'onlineSavingsMsg'
Learn More

(±)-PPCC oxalate, Tocris Bioscience™

Product Code. 18065902
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective sigma (σ) agonist (σ1> σ2)

Selective sigma (σ) receptor ligand. Displays high affinity for σ1; also binds at σ2 sites (Ki = 1.5 nM and 50.8 nM respectively). Selective over a range of receptor types including dopaminergic and muscarinic receptors, DAT and SERT.

Chemical Identifiers

CAS 932736-91-9
Molecular Formula C52H62N2O14
Molecular Weight (g/mol) 939.068
InChI Key IGJDSEGFEVKASG-KLELIYFTSA-N
Synonym (S™, R™)-2-[(4-Hydroxy-4-phenyl-1-piperidinyl)methyl]-1-(4-methylphenyl)-cyclopropanecarboxylic acid methyl ester
PubChem CID 90488889
IUPAC Name methyl (1S,2R)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;methyl (1R,2S)-2-[(4-hydroxy-4-phenylpiperidin-1-yl)methyl]-1-(4-methylphenyl)cyclopropane-1-carboxylate;oxalic acid
SMILES CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.CC1=CC=C(C=C1)C2(CC2CN3CCC(CC3)(C4=CC=CC=C4)O)C(=O)OC.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 469.53
Percent Purity >99%
Chemical Name or Material (±)-PPCC oxalate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'