missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPNDS, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective P2X1 receptor antagonist (pKB = 7.43 in rat vas deferens). Up to 52-fold selective over P2Y1 receptors, and selective over ecto-nucleotidase, α1A-adrenoceptors, A1, A2B, H1 and M3 receptors.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 694.36 |
Percent Purity | >98% |
Chemical Name or Material | PPNDS |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'