missing translation for 'onlineSavingsMsg'
Learn More

PPNDS, Tocris Bioscience™

Product Code. 18094622
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective P2X1 antagonist

Potent and selective P2X1 receptor antagonist (pKB = 7.43 in rat vas deferens). Up to 52-fold selective over P2Y1 receptors, and selective over ecto-nucleotidase, α1A-adrenoceptors, A1, A2B, H1 and M3 receptors.

Chemical Identifiers

CAS 1021868-77-8
Molecular Formula C18H11N4Na4O14PS2
Molecular Weight (g/mol) 694.353
InChI Key TXFLDKHOZFWVAC-UHFFFAOYSA-J
Synonym ppnds tetrasodium
PubChem CID 57369378
IUPAC Name tetrasodium;3-[2-[4-formyl-6-methyl-5-oxo-3-(phosphonatooxymethyl)pyridin-2-ylidene]hydrazinyl]-7-nitronaphthalene-1,5-disulfonate
SMILES CC1=NC(=NNC2=CC(=C3C=C(C=C(C3=C2)S(=O)(=O)[O-])[N+](=O)[O-])S(=O)(=O)[O-])C(=C(C1=O)C=O)COP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 50 mg
Formula Weight 694.36
Percent Purity >98%
Chemical Name or Material PPNDS
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'