missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R9B Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA548102
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
PPP1R9B Polyclonal antibody specifically detects PPP1R9B in Human, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
PPP1R9B | |
Polyclonal | |
Unconjugated | |
Ppp1r9b | |
Neb2; neurabin 2; neurabin II; neurabin-2; Neurabin-II; neural tissue-specific F-actin-binding protein II; nuerabin 2; p130; PP1bp134; PPP1R6; PPP1R9; PPP1R9B; Protein phosphatase 1 regulatory subunit 9B; protein phosphatase 1, regulatory (inhibitor) subunit 9B; protein phosphatase 1, regulatory subunit 9B; protein phosphatase 1, regulatory subunit 9B, spinophilin; SPINO; Spinophilin; SPL; Spn | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
84686, 84687 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35274, Q96SB3 | |
Ppp1r9b | |
E. coli-derived recombinant human PPP1R9B Lys289-Asp412 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |