missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPT, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, subtype-selective estrogen receptor agonist (EC50 ∽ 200 pM); displays 410-fold selectivity for ERα over ERβ. Prevents ovariectomy-induced weight gain and loss of bone mineral density, and induces gene expression in the hypothalamus following systemic administration in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 386.45 |
Percent Purity | >99% |
Chemical Name or Material | PPT |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'