missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10887-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene.Specifications
PPT1 | |
Polyclonal | |
Unconjugated | |
PPT1 | |
CLN1, INCL, Palmitoyl protein hydrolase 1, PPT, PPT 1, PPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19063, 29411, 5538 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88531, P45479, P50897 | |
Ppt1 | |
PPT1 Fusion Protein Ag1131 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction