missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548700
302.40 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
This gene encodes a member of the palmitoyl-protein thioesterase family. The encoded glycosylated lysosomal protein has palmitoyl-CoA hydrolase activity in vitro, but does not hydrolyze palmitate from cysteine residues in proteins. Three transcript variants encoding different isoforms have been described for this gene, with one of the isoforms being inactive.
Specifications
PPT2 | |
Polyclonal | |
Unconjugated | |
PPT2 | |
0610007M19Rik; AA672937; C6orf8; G14; Lysosomal thioesterase PPT2; palmitoyl-protein hydrolase 2; palmitoyl-protein thioesterase 2; PPT2; PPT-2; S-thioesterase G14; VE-statin2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
9374 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q9UMR5 | |
PPT2 | |
KLH conjugated synthetic peptide between 183-212 amino acids from the Central region of human PPT2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PPT2 Polyclonal Antibody, Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction