Learn More
Atago™ PR-301α Model Digital Refractometer
Atago™ PR-301α Model Digital Refractometer is designed to cover high concentration of Brix 45.0 to 90.0%.
Brand:  Atago™ 3462
Product Code. 10779044
  • £2000.00 / Each
Explore more special offers
Description
Description
  • Digital refractometer
  • Improved measurement accuracy
  • Compact and battery operated
  • Display the measured values of samples quickly
Specifications
Specifications
Digital Refractometer
PR-301α
0.1% Brix
The measurements of jellies, raw liquid sugar
IP64
10°C to 40°C
45 to 90 Brix
±0.1% Brix at 10 to 30°C
Brix
Standard liquid LG, Round sheet
300 g
006P dry battery (9V)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products