missing translation for 'onlineSavingsMsg'
Learn More

Praseodymium(III) carbonate octahydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 11342568
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
14948-62-0
C3H16O17Pr2
605.959
MFCD00211307
SEHQEMTYQGRHIC-UHFFFAOYSA-H
praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate
25022096
praseodymium(3+);tricarbonate;octahydrate
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3]
This item is not returnable. View return policy
14948-62-0
C3H16O17Pr2
605.959
MFCD00211307
SEHQEMTYQGRHIC-UHFFFAOYSA-H
praseodymium carbonate octahydrate, 3co3.2pr.8h2o, praseodymium carbonate, praseodymium carbonate-water 2/3/8, dipraseodymium 3+ ion octahydrate trico3, praseodymium iii carbonate octahydrate, reacton, dipraseodymium 3+ octahydrate tricarbonate
25022096
praseodymium(3+);tricarbonate;octahydrate
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.O.O.O.O.[Pr+3].[Pr+3]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Green
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.9% (REO)
Formula Weight 605.97 (461.85 Anhydrous)
Odor Odorless
Appearance Green crystalline
Chemical Name or Material Praseodymium(III) carbonate octahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'