Learn More
Praseodymium(III, IV) oxide, 99.9% (REO), Thermo Scientific Chemicals
£55.98 - £173.87
Chemical Identifiers
CAS | 12037-29-5 |
---|---|
Molecular Formula | O11Pr6 |
Molecular Weight (g/mol) | 1021.44 |
MDL Number | MFCD00011178 |
InChI Key | HPZIIFVSYNLWNX-UHFFFAOYSA-N |
IUPAC Name | hexapraseodymium(3+) undecaoxidandiide |
SMILES | [O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323348
|
Thermo Scientific Alfa Aesar
036276.09 |
10 g |
£55.98
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11313348
|
Thermo Scientific Alfa Aesar
036276.18 |
50 g |
£173.87
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Praseodymium(III, IV) oxide is used in pigment in glass, optic and ceramic industries. Praseodymium-doped glass, called didymium glass is used in welding, blacksmithing, and glass-blowing goggles due to its blocking property of infrared radiation. It is employed in the solid state synthesis of praseodymium molybdenum oxide, which is used as a semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12037-29-5 | |
1021.44 | |
HPZIIFVSYNLWNX-UHFFFAOYSA-N | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3]Show More |
O11Pr6 | |
MFCD00011178 | |
hexapraseodymium(3+) undecaoxidandiide |
Specifications
2500°C | |
Sintered Lump | |
MFCD00011178 | |
HPZIIFVSYNLWNX-UHFFFAOYSA-N | |
hexapraseodymium(3+) undecaoxidandiide | |
1021.43 | |
(REO) |
12037-29-5 | |
O11Pr6 | |
Insoluble in water. Moderately soluble in strong mineral acids. | |
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3].[Pr+3] | |
1021.44 | |
99.9% | |
Praseodymium(III, IV) oxide |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 234-857-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Praseodymium(III, IV) oxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Praseodymium(III, IV) oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.